Learn More
StatLab MasterTech Steiner Stain Kit
SDP
Catalog No. 21033408
Change view
Click to view available options
Volume (Metric):
100 mL
Catalog No. Volume (Metric)
21-033-408 100 mL
1 options
Catalog No. 21-033-408 Supplier StatLab Supplier No. KTSTE
Add to Cart
Add to Cart
Stains for donovan bodies, fungi, helicobacter pylori, legionella pneumophila, and spirochetes in 45 minutes
  • The MasterTech Steiner Stain Kit differs from similar kits by eliminating radioactive Uranyl Nitrate, providing pre-measured Hydroquinone capsules, and modifying the original technique, which creates a safer, easier to use procedure that provides optimal staining results and reduced background staining
  • Silver Nitrate, developed by the Hydroquinone based reducing solution serves as the primary stain, producing dark brown to black Donovan bodies, fungi, Helicobacter pylori, legionella pneumophila, and spirochetes, against a yellow to brown background
  • 100 mL Stain Kit yields between 20-32 slides
  • 100 mL Kit Consists of: 10% Zinc Formalin 100 mL, 1% Silver Nitrate 5 mL, 0.1% Silver Nitrate 100 mL, Hydroquinone Capsules 5 each, Oxidizer Solution 100 mL and 2.5% Gum Mastic 125 mL
  • A MasterTech Special Stain Kit
Specifications
Volume (Metric) 100 mL
Product Type Steiner Stain Kit