Learn More
MasterTech Toluidine Blue Stock Solution, StatLab
SDP
Catalog No. 22024789
Change view
Click to view available options
Quantity:
10 mL
Catalog No. Quantity
22-024-789 10 mL
1 options
Catalog No. 22-024-789 Supplier SLMP Llc Dba Statlab Medical Supplier No. KC372J
Add to Cart
Add to Cart
Used to selectively stain acidic tissue components.
  • A cationic (basic) thiazine dye used to selectively stain acidic tissue components
  • It is highly regarded for its metachromatic properties, meaning it can stain certain tissue components a different color (purple/red) than the dye solution itself (blue)
  • A StatLab MasterTech Special Stain Kit Component
Specifications
Quantity 10 mL