Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Toluidine Blue Stock Solution
SDP

Catalog No. 21033383
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-383 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-383 Supplier StatLab Supplier No. KC3721
Add to Cart
Add to Cart

Used to identify and highlight specific tissue components like mast cells, cartilage, and mucin

  • Used to identify and highlight specific tissue components like mast cells, cartilage, and mucin due to its metachromatic staining properties where it binds strongly to acidic substances within the tissue causing them to appear a different color than the dye itself
  • A component for the MasterTech HelicoStat Rapid Helicobacter Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material Toluidine Blue
Grade Laboratory
Color Blue
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.