Learn More
StatLab MasterTech TransForm Complete Neutralizing Dry-Crystals
SDP
Catalog No. 21010284
Change view
Click to view available options
Material:
Crystalline Chemical Mixture
Catalog No. Material
21-010-284 Crystalline Chemical Mixture
1 options
Catalog No. 21-010-284 Supplier StatLab Supplier No. CWTFRCS
Add to Cart
Add to Cart
The latest advancement in 10% Formalin Neutralization.
  • Each easy-to-use packet will neutralize one gallon of 10% Formalin into a non-hazardous, neutral pH clear-liquid, in minutes
  • EPA test results are attached to product information sheet for quick reference
  • TransForm Complete requires no hazardous shipping charge and is inexpensive to ship (weighs only 17 pounds)
  • it complies with State and Federal waste-water regulations
  • Each case neutralizes 10 gallons of 10% Formalin
Specifications
Material Crystalline Chemical Mixture
Type Formalin Neutralizer
Packaging Quantity 10 Pk.