Learn More
StatLab™ MasterTech Trichrome Masson's Stain Kit
SDP
Catalog No. STLKTMTR
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLKTMTR 100 mL
STLKTMTRPT 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTMTR Supplier StatLab™ Supplier No. KTMTR
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Masson's Stain Kit stains for collagen and muscle in as little 20 minutes.
  • Biebrich Scarlet/Acid Fuchsin and Aniline Blue serve as the primary stains, producing red cytoplasm, intercellular fiber, keratin, and muscle, blue collagen and mucus with purple to black nuclei; counterstained by Weigert's Hematoxylin
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Quantity 100 mL
Product Type Stain Kit