Learn More
StatLab MasterTech Trichrome Special Stain Kit - Masson's
SDP
Catalog No. 21010340
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
21-010-340 1 L
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-340 Supplier StatLab Supplier No. KTMTRLT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Stains for collagen and muscle in as little 20 min.
  • Trichrome stains are commonly used to identify increases in collagenous tissue to assist in the diagnosis of diseases such as liver cirrhosis
  • This kit includes a 20 min. microwave procedure and the traditional one hour and forty-five minute procedure
  • Biebrich Scarlet/Acid Fuchsin and Aniline Blue serve as the primary stains, producing red cytoplasm, intercellular fiber, keratin, and muscle, blue collagen and mucus with purple to black nuclei, counterstained by Weigert’s Hematoxylin
  • Liter Kit Consists of: Bouins Fluid Liter, Biebrich Scarlet/Acid Fucshin Liter, Phosphomolybdic/Phoshotungstic Acid Liter, Aniline Blue Stain Liter, 1% Acetic Acid Liter and Weigert's A and B Hematoxylin 2 Pints each
  • Liter Stain Kit yields between 680-1,000 slides.
  • A MasterTech Special Stain Kit
Specifications
Quantity 1 L
Product Type Masson's Trichrome