Learn More
StatLab MasterTech Trichrome Stain Kit - One-Step Blue and Red
SDP
Catalog No. 21033409
Change view
Click to view available options
Quantity:
16 oz.
Catalog No. Quantity
21-033-409 16 oz.
1 options
Catalog No. 21-033-409 Supplier StatLab Supplier No. KTTRBPT
Add to Cart
Add to Cart
Stains for collagen and muscle in one hour and 15 minutes
  • The MasterTech One-Step Trichrome Kit makes staining for collagen or muscle, fast and easy
  • Unlike traditional one-step procedures, Modified Mayers Hematoxylin is used instead of Weigert's Hematoxylin, eliminating the need for unnecessary steps, while making this kit ready-to-use
  • One Step Trichrome serves as the primary stain, producing pink to red cytoplasm, intercellular fiber, keratin, and muscle, blue or green collagen, and mucus, depending on the Trichrome color chosen, and blue nuclei stained by Mayers Hematoxylin
  • Pint Blue/Red Stain Kit yields between 320-500 slides
  • One-Step Blue/Red Pint Kit Consists of: Bouins Fluid Pint, Modified Mayers Hematoxylin Pint, One Step Trichrome Blue/Red Pint
  • A MasterTech Special Stain Kit
Specifications
Quantity 16 oz.
Product Type Masson's Trichrome