Learn More
StatLab MasterTech Trichrome Stain Kit, One-Step Green and Red

Description
- The MasterTech One-Step Trichrome Kit makes staining for collagen or muscle fast and easy
- Unlike traditional one-step procedures Modified Mayers Hematoxylin is used instead of Weigerts Hematoxylin eliminating the need for unnecessary steps while making this kit ready-to-use
- One Step Trichrome serves as the primary stain producing pink to red cytoplasm intercellular fiber keratin and muscle blue or green collagen and mucus depending on the Trichrome color chosen and blue nuclei stained by Mayer's Hematoxylin
- A MasterTech Special Stain Kit
Specifications
Specifications
| Color | Pink to Red |
| Quantity | 1 pt. |
| Grade | Histological |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.