Learn More
StatLab MasterTech Universal Iodine Solution
SDP
Catalog No. 21033468
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
21-033-468 100 mL
1 options
Catalog No. 21-033-468 Supplier StatLab Supplier No. STUIO100
Add to Cart
Add to Cart
Used to visualize and study cell structures in greater detail
  • It is used in a variety of applications, including microbiology, pathology, and plant biology
  • Without iodine solution, many of the components of a specimen would be invisible under the microscope
  • Used in the Brown and Benn, Elastic, Gram and Pentachrome stain kits
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 100 mL
Product Type Universal Iodine