Learn More
StatLab™ MasterTech Verhoeff's Elastic Stain Kit
SDP
Catalog No. STLKTVELPT
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLKTVELPT 1 pt.
STLKTVEL 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTVELPT Supplier StatLab™ Supplier No. KTVELPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Elastic Stain Kit stains for elastic fibers in just 20 minutes.
  • Verhoeff's working solution serves as the primary stain in this regressive method, overstained and then differentiated to produce purple to black elastic fibers and pink to red collagen against yellow background tissue counterstained by Van Gieson's
  • 100 mL Stain Kit yields between 36 and 45 slides
  • 1 pt. Stain Kit yields between 144 and 225 slides
Specifications
Quantity 1 pt.
Product Type Stain Kit