Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Von Kossa Stain Kit
SDP

Catalog No. 21033410
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-410 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-410 Supplier StatLab Supplier No. KTVKOPT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stains for calcium deposits in just 20 minutes

  • Silver Nitrate serves as the primary stain, reacting with the anions and ultra-violet light to produce brown to black calcium salts against pink to red cytoplasm and nuclei, counterstained by Nuclear Fast Red
  • There is no filtration required when using MasterTech Nuclear Fast Red
  • Pint Stain Kit yields between 50-192 slides
  • Pint Kit Consists of: 5% Silver Nitrate pint, 5% Sodium Thiosulfate pint and Nuclear Fast Red Stain pint
  • A MasterTech Special Stain Kit

Specifications

Quantity 16 oz.
Product Type Von Kassa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.