Learn More
StatLab MasterTech Warthin-Starry Stain Kit - Modified Stat
SDP
Catalog No. 21033411
Change view
Click to view available options
Quantity:
16 oz.
Catalog No. Quantity
21-033-411 16 oz.
1 options
Catalog No. 21-033-411 Supplier StatLab Supplier No. KTWSTPT
Add to Cart
Add to Cart
Stains cat scratch fever, H. pylori, l. pneumophila, and spirochetes in 15 minutes
  • This kit is suitable in lieu of and comparable to the traditional Warthin-Starry Stain
  • This kit differs from the traditional procedure by incorporating optimized reagents which allows the MasterTech Warthin-Starry Staining time to be reduced to just 15 minutes instead of more than two hours
  • Silver Nitrate, developed by the Hydroquinone based reducing solution serves as the primary stain, producing dark brown to black cat scratch fever, Helicobacter pylori, legionella pneumophila, and spirochetes against a yellow to brown background
  • Pint Stain Kit yields between 50-256 slides
  • Pint Kit Consists of: 2% Silver Nitrate Acidulated 100 mL, 0.5% Silver Nitrate Acidulated pint, 4% Gelatin Acidulated 400 mL, 0.1% Hydroquinone Acidulated 400ml, Spirochete Sensitizer Solution pint
  • A MasterTech Special Stain Kit
Specifications
Quantity 16 oz.
Product Type Warthin-Starry