Learn More
StatLab MasterTech Weigert's Hematoxylin
Description
- Used in histology to stain nuclei, particularly when a subsequent staining step in the protocol involves an acidic solution
- Highly resistant to decolorization by acids, making it ideal for counterstaining in special staining techniques where other hematoxylin stains might be washed out
Specifications
Specifications
| Product Type | Weigert's Hematoxylin B |