Learn More
StatLab MasterTech Weigert's Hematoxylin A
SDP
Catalog No. 21010298
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
21-010-298 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-298 Supplier StatLab Supplier No. HXWHEA100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Used for staining nuclei in tissues.
  • Combine equal parts of A and B MasterTech Weigerts Hematoxylin
  • It is particularly useful when acid solutions are used in a later step of the staining protocol
  • The hematoxylin-eosin staining is one of the most widely used techniques in histology labs, and Weigert's Hematoxylin is part of the van Gieson's trichrome staining procedure
  • This staining method is effective for visualizing the chromatin of the nucleus and is often used in conjunction with other staining techniques to provide a comprehensive view of the tissue sample
Specifications
Quantity 100 mL