Learn More
StatLab MasterTech Weigert's Hematoxylin B
SDP
Catalog No. 21060043
Change view
Click to view available options
Quantity:
100 mL
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-060-043 100 mL
21-060-019 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-060-043 Supplier StatLab Supplier No. HXWHEB100
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Used for staining nuclei in tissues.
  • Combine equal parts of A and B MasterTech Weigerts Hematoxylin
  • It is particularly useful when acid solutions are used in a later step of the staining protocol
  • The hematoxylin-eosin staining is one of the most widely used techniques in histology labs, and Weigert's Hematoxylin is part of the van Gieson's trichrome staining procedure
  • This staining method is effective for visualizing the chromatin of the nucleus and is often used in conjunction with other staining techniques to provide a comprehensive view of the tissue sample
Specifications
Quantity 100 mL