Learn More
StatLab MasterTech Weigert's Hematoxylin B
SDP
Catalog No. 22021383
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
22-021-383 1 L
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-383 Supplier StatLab Supplier No. HXWHEBLT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Weigert's Hematoxylin B - Combine equal parts of A and B MasterTech Weigerts Hematoxylin
  • Used primarily for nuclear staining particularly when a staining procedure involves acidic solutions due to its high resistance to decolorization under acidic conditions
Specifications
Quantity 1 L
Product Type Weigert's Iron Hematoxylin- Part B