Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Xylene Peanut Oil
SDP

Catalog No. 22024788
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-024-788 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-024-788 Supplier StatLab Supplier No. KC2542
Add to Cart
Add to Cart

Used to detect Leprosy and Nocardia in Staining procedure.

  • It acts as a gentle deparaffinizing agent, where peanut oil protects delicate, acid-fast bacterial cell walls from being damaged by harsh solvent effects of pure xylene during staining
  • Offers a gentler alternative to pure xylene for delicate tissues, acting as both a cleanser and a protective coating
  • A StatLab MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
Color Yellow
Quantity 500 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.