Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Digital Drive with Multichannel Cartridge Pump Head for L/S Precision Tubing, 4-Channel, 8-Roller, 100 rpm; 115/230 VAC
Catalog No. NC1721839
Encompass
These precision drives offer analog remote control options and simple programming¿ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy¿drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
Catalog No. NC1721839 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7791935
Add to Cart
Add to Cart
Request a Quote
These precision drives offer analog remote control options and simple programming¿ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy¿drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.