Learn More
Sino Biological Recombinant Human MYO3B Protein (GST Tag), Active
Catalog No. 502574105
Encompass_Preferred
Recombinant human MYO3beta (1-326) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4105 Supplier Sino Biological Supplier No. M6611G50
Recombinant human MYO3beta (1-326) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.