Learn More
Aviva Systems Biology Corp MASTL Antibody - C-terminal region (ARP60688_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60688_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MASTL. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MASTL. Alternate names: FLJ14813, RP11-85G18.2, THC2, GW, GWL, hGWL, MAST-L, GREATWALLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-126