Learn More
Medchemexpress LLC MASTL-IN-4 | 2883041-36-7 | 99.7% | 353.33 | 1 MG
SDP
Catalog No. 5000392705
Encompass_Preferred
MASTL-IN-4 (compound 70) is a potent MASTL inhibitor with a pIC50 value of 9.15. It demonstrates anti-proliferative activity. It is for research use only and not sold to patients.
  • Potent MASTL inhibitor with a pIC50 value of 9.15
  • Shows anti-proliferative activity
  • Available in various quantities
  • Stored at 4°C under nitrogen, or in solvent at -80°C for 6 months; -20°C for 1 month (under nitrogen)
Catalog No. 50-003-92705 Supplier Medchemexpress LLC Supplier No. HY1627321MG
Add to Cart
Add to Cart
MASTL-IN-4 (compound 70) is a potent MASTL inhibitor with a pIC50 value of 9.15. It demonstrates anti-proliferative activity. It is for research use only and not sold to patients.
  • Potent MASTL inhibitor with a pIC50 value of 9.15
  • Shows anti-proliferative activity
  • Available in various quantities
  • Stored at 4°C under nitrogen, or in solvent at -80°C for 6 months; -20°C for 1 month (under nitrogen)