Learn More
Medchemexpress LLC MASTL Kinase Inhibitor-1 | 1190277-80-5 | 98.1% | 302.33 | 50 MG
SDP
Catalog No. 5000349000
Encompass_Preferred
MKI-1 is an inhibitor of MASTL (microtubule-associated serine/threonine kinase-like) with an IC50 of 9.9 μM. It exhibits antitumor and radiosensitizer activities through PP2A activation in breast cancer.
  • Inhibits MASTL activity in breast cancer cells (5-20 μM)
  • Inhibits oncogenic properties of breast cancer cells at 100 μM, with weaker effects on normal breast cells
  • Reduces serine 62-phosphorylation of c-Myc and total c-Myc, with a decrease in ENSA phosphorylation
  • Reduces c-Myc stability through PP2A activation in MCF7 cells
  • Reduces tumor growth and enhances radiosensitivity in BT549 xenograft model (50 mg/kg, ip, twice a week)
Catalog No. 50-003-49000 Supplier Medchemexpress LLC Supplier No. HY13755250MG
MKI-1 is an inhibitor of MASTL (microtubule-associated serine/threonine kinase-like) with an IC50 of 9.9 μM. It exhibits antitumor and radiosensitizer activities through PP2A activation in breast cancer.
  • Inhibits MASTL activity in breast cancer cells (5-20 μM)
  • Inhibits oncogenic properties of breast cancer cells at 100 μM, with weaker effects on normal breast cells
  • Reduces serine 62-phosphorylation of c-Myc and total c-Myc, with a decrease in ENSA phosphorylation
  • Reduces c-Myc stability through PP2A activation in MCF7 cells
  • Reduces tumor growth and enhances radiosensitivity in BT549 xenograft model (50 mg/kg, ip, twice a week)