Learn More
Cayman Chemical Mastoparn 7trIfluoroACTAt 10mg
SDP
Catalog No. 502942338
Encompass_Preferred
A G protein-activating peptide; decreases the affinity of ACh to porcine brain mAChRs at 30 µM; induces activation of Gαo in, and increases dendritic spine density of, primary rat hippocampal neurons at 1 µM; induces contractions in isolated perfused rat tail arteries (EC50 = 44.1 nM); immunization with a vaccine against cocaine in combination with mastoparan 7 as an adjuvant increases hapten-specific serum IgG levels and decreases cocaine-induced locomotion in mice
Catalog No. 50-294-2338 Supplier Cayman Chemical Supplier No. 4011510 MG
A G protein-activating peptide; decreases the affinity of ACh to porcine brain mAChRs at 30 µM; induces activation of Gαo in, and increases dendritic spine density of, primary rat hippocampal neurons at 1 µM; induces contractions in isolated perfused rat tail arteries (EC50 = 44.1 nM); immunization with a vaccine against cocaine in combination with mastoparan 7 as an adjuvant increases hapten-specific serum IgG levels and decreases cocaine-induced locomotion in mice