Learn More
TARGETMOL CHEMICALS INC MASUPIRDINE FREE BASE 5MG
SDP
Catalog No. 502577435
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. Masupirdine free base (SUVN-502 free base) is an orally available 5-HT6 receptor antagonist that crosses the blood-brain barrier and has a Ki value of 2.04 nM for the human 5-HT6 receptor.Masupirdine free base is potent and selective with multiple targets. Masupirdine free base is highly potent selective and inhibits multiple targets.Masupirdine free base can be used in the study of neurological disorders such as Alzheimer's disease. purity: 98%
Catalog No. 50-257-7435 Supplier TARGETMOL CHEMICALS INC Supplier No. T119485MG
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. Masupirdine free base (SUVN-502 free base) is an orally available 5-HT6 receptor antagonist that crosses the blood-brain barrier and has a Ki value of 2.04 nM for the human 5-HT6 receptor.Masupirdine free base is potent and selective with multiple targets. Masupirdine free base is highly potent selective and inhibits multiple targets.Masupirdine free base can be used in the study of neurological disorders such as Alzheimer's disease. purity: 98%