Learn More
TARGETMOL CHEMICALS INC Masupirdine mesylate 10MG
SDP
Catalog No. 502879860
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Masupirdine mesylate (SUVN-502 mesylate) is a selective 5-HT6 receptor antagonist that crosses the blood-brain barrier and is utilized in the study of neurological disorders such as Alzheimer's disease and dementia. Purity 99.48%
Catalog No. 50-287-9860 Supplier TARGETMOL CHEMICALS INC Supplier No. T1194910MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Masupirdine mesylate (SUVN-502 mesylate) is a selective 5-HT6 receptor antagonist that crosses the blood-brain barrier and is utilized in the study of neurological disorders such as Alzheimer's disease and dementia. Purity 99.48%