Learn More
ACL Inc MAT ANTI-STATIC ESD DUALMAT
Catalog No. 501037927
Encompass_Preferred
ESD Workbench Products, Anti-Static Mats, Static Dissipative Mats, Staticide Dualmat, 24 in.x72 in. dark gray pre-cut mat, 2-layer rubber, 10^6-10^8 ohms per square, compliant to ANSI / ESDA S20.20
Catalog No. 50-103-7927 Supplier ACL Inc Supplier No. 8385DGYM2472
ESD Workbench Products, Anti-Static Mats, Static Dissipative Mats, Staticide Dualmat, 24 in.x72 in. dark gray pre-cut mat, 2-layer rubber, 10^6-10^8 ohms per square, compliant to ANSI / ESDA S20.20