Learn More
ACL Inc MAT ANTI-STATIC ESD SPECMAT-H
Catalog No. 501037912
Encompass_Preferred
ESD Workbench Products, Anti-Static Mats, Static Dissipative Mats, Staticide Spec-Mat H, 24 in.x36 in. light blue pre-cut mat, single layer homogeneous thermoplastic vinyl, 10^8-10^9 ohms per square, compliant to ANSI / ESDA S20.20. Mat thickness .100 in.
Catalog No. 50-103-7912 Supplier ACL Inc Supplier No. 6212436
ESD Workbench Products, Anti-Static Mats, Static Dissipative Mats, Staticide Spec-Mat H, 24 in.x36 in. light blue pre-cut mat, single layer homogeneous thermoplastic vinyl, 10^8-10^9 ohms per square, compliant to ANSI / ESDA S20.20. Mat thickness .100 in.