Learn More
Chemglass Life Sciences Mat for 96-Well PCR Plates, Blue, Santoprene
SDP
Catalog No. 501311313
Encompass_Preferred
Mat for 96-Well PCR Plates, Blue, Santoprene. This flexible sealing cover is used on 96-well plates along with clip down and screw top thermal cyclers and has been proven to be a secure and effective way of sealing. Since it is reusable, it is a nice way to make this step of the procedure cost effective. Dimples on one side of the mat ensure it is well placed over the tubes. Autoclavable.
Catalog No. 50-131-1313 Supplier Chemglass Life Sciences Supplier No. CLS360901
Mat for 96-Well PCR Plates, Blue, Santoprene. This flexible sealing cover is used on 96-well plates along with clip down and screw top thermal cyclers and has been proven to be a secure and effective way of sealing. Since it is reusable, it is a nice way to make this step of the procedure cost effective. Dimples on one side of the mat ensure it is well placed over the tubes. Autoclavable.