Learn More
Medchemexpress LLC HY-112569 5mg Medchemexpress, MAT2A inhibitor 2 CAS:13299-99-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1125695MG
Encompass_Preferred
Medchemexpress, HY-112569 5mg MAT2A inhibitor 2 CAS:13299-99-5 MAT2A inhibitor 2 is a methionine adenosyltransferase 2A (MAT2A) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2424