Learn More
Aviva Systems Biology Corp MAT2B Antibody - middle region (ARP55850_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55850_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAT2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAT2B. Alternate names: MAT-II, MATIIbeta, MGC12237, Nbla02999, TGR, SDR23E1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61634