Learn More
Cayman Chemical Matairesinol Fatty Acids and D
SDP
Catalog No. 502888768
Encompass_Preferred
A lignan with diverse biological activities; reduces LPS-induced production of NO and PGE2 in RAW 264.7 cells; cytotoxic to colon 26 cancer cells (LC50 = 9 UG/ml); increases the lifespan of C. elegans at 100 µM; reduces oxaliplatin-induced neurotoxicity in neural PC12 cells at 0.4, 2, and 10 µM; topical administration reduces EAr thickness and mast cell infiltration, as well as serum IgE levels, in a mouse model of dust mite-induced atopic dermatitis at 50 UG/ear
Catalog No. 50-288-8768 Supplier Cayman Chemical Supplier No. 1000517410 MG
A lignan with diverse biological activities; reduces LPS-induced production of NO and PGE2 in RAW 264.7 cells; cytotoxic to colon 26 cancer cells (LC50 = 9 UG/ml); increases the lifespan of C. elegans at 100 µM; reduces oxaliplatin-induced neurotoxicity in neural PC12 cells at 0.4, 2, and 10 µM; topical administration reduces EAr thickness and mast cell infiltration, as well as serum IgE levels, in a mouse model of dust mite-induced atopic dermatitis at 50 UG/ear