Learn More
Cayman Chemical Matairesinol, Purity: ≥95%, CAS: 580-72-3, 5 mg
SDP
Catalog No. 502889286
Encompass_Preferred
A lignan with diverse biological activities; reduces LPS-induced production of NO and PGE2 in RAW 264.7 cells; cytotoxic to colon 26 cancer cells (LC50 = 9 µg/ml); increases the lifespan of C. elegans at 100 µM; reduces oxaliplatin-induced neurotoxicity in neural PC12 cells at 0.4, 2, and 10 µM; topical administration reduces ear thickness and mast cell infiltration, as well as serum IgE levels, in a mouse model of dust mite-induced atopic dermatitis at 50 µg/ear
Catalog No. 50-288-9286 Supplier Cayman Chemical Supplier No. 100051745 MG
A lignan with diverse biological activities; reduces LPS-induced production of NO and PGE2 in RAW 264.7 cells; cytotoxic to colon 26 cancer cells (LC50 = 9 µg/ml); increases the lifespan of C. elegans at 100 µM; reduces oxaliplatin-induced neurotoxicity in neural PC12 cells at 0.4, 2, and 10 µM; topical administration reduces ear thickness and mast cell infiltration, as well as serum IgE levels, in a mouse model of dust mite-induced atopic dermatitis at 50 µg/ear