Learn More
HCL Labels Match Arrows To Ensure Adequate Ventilation - 3.5"""" x 5"""" Adhesive Vinyl Label
SDP
Catalog No. NC1490905
Encompass_Preferred
Match Arrows To Ensure Adequate Ventilation - 3.5"""" x 5"""" Adhesive VinylMatch Arrows To Ensure Adequate Ventilation Fume Hood Label - two part label, both parts are adhesive, and offer flexibility of placement.This label is used on a chemical fume hood. It is in two parts to ensure that once the labels are placed, the user has adequate ventilation when operating the fume hood.
Catalog No. NC1490905 Supplier HCL Labels Supplier No. SSL0012
Match Arrows To Ensure Adequate Ventilation - 3.5"""" x 5"""" Adhesive VinylMatch Arrows To Ensure Adequate Ventilation Fume Hood Label - two part label, both parts are adhesive, and offer flexibility of placement.This label is used on a chemical fume hood. It is in two parts to ensure that once the labels are placed, the user has adequate ventilation when operating the fume hood.