Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quanterix Inc MATCHED LOT HELPER BEADS
SDP

Catalog No. NC1485349
Encompass

  • Critical component for developing sensitive assays.
  • Designed for use with Simoa HD-1, HD-X, and SR-X instruments.
  • Enables ultrasensitive detection and quantification of proteins.
  • Non-biologic, non-toxic, and non-infectious mixture.
  • Intended strictly for laboratory research use.
  • Matched lot characteristic ensures consistency and reliability.
  • Made of carboxylated paramagnetic beads, dye-encoded.
  • Bead size is 2.7 µm.
  • Volume of 1.5 mL.
  • Concentration of 1.25x109 Beads/mL.
  • Storage temperature range of 2-8°C.
  • Liquid suspension format suitable for immunoassay.
  • Key composition includes 90-100% water and 0.1-1% sodium chloride.

Catalog No. NC1485349 Supplier Quanterix Inc Supplier No. 103208
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Critical component for developing sensitive assays.
  • Designed for use with Simoa HD-1, HD-X, and SR-X instruments.
  • Enables ultrasensitive detection and quantification of proteins.
  • Non-biologic, non-toxic, and non-infectious mixture.
  • Intended strictly for laboratory research use.
  • Matched lot characteristic ensures consistency and reliability.
  • Made of carboxylated paramagnetic beads, dye-encoded.
  • Bead size is 2.7 µm.
  • Volume of 1.5 mL.
  • Concentration of 1.25x109 Beads/mL.
  • Storage temperature range of 2-8°C.
  • Liquid suspension format suitable for immunoassay.
  • Key composition includes 90-100% water and 0.1-1% sodium chloride.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.