Learn More
Biochain Institute Inc Matched Pair - Total Protein - Human Primary Tumor and Normal Tissue (PP): Colon, 0.2 x2 mg /PK
SDP
Catalog No. 50181702
Encompass_Preferred
BioChain's human total protein matched pair products include: Primary Pair (PP), Primary and Metastatic Pair (PM). PP consists of total protein isolated from primary tumor and its adjacent normal tissue; PM consists of total protein from primary tumor andcorresponding metastastic tumor. Total proteins in each pair are prepared from the same donor. This product line is designedfor identifying tumor-specific genes and tumor metastasis genes.
Catalog No. 50-181-702 Supplier Biochain Institute Inc Supplier No. P8235090PP
BioChain's human total protein matched pair products include: Primary Pair (PP), Primary and Metastatic Pair (PM). PP consists of total protein isolated from primary tumor and its adjacent normal tissue; PM consists of total protein from primary tumor andcorresponding metastastic tumor. Total proteins in each pair are prepared from the same donor. This product line is designedfor identifying tumor-specific genes and tumor metastasis genes.