Learn More
Biochain Institute Inc Matched Pair - Frozen Tissue Section - Human Primary Tumor and Normal (PP): Breast, 5 x2 slides /PK
SDP
Supplier:  Biochain Institute Inc T6235086PP
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the ethical standard and IRB-approved procedures. During the collection, the excised tissues are immediately put into liquid nitrogen and then identified by a board-licensed pathologist. Patient demographic and disease related history data are available. Human frozen tissue section matched pair products include: Primary Pair (PP), Primary and Metastatic Pair (PM). PP consists of frozen tissue sections of primary tumor and its adjacent normal tissue; PM consists of frozen tissue sections of primary tumor and corresponding metastatic tumor. Frozen tissue sections in each pair are prepared from the same donor. This product line is ideal for localization of tumor-specific genes and tumor metastatic genes.
Catalog No. 50-181-672