Learn More
Biochain Institute Inc Matched Pair - Total RNA - Human Primary and Metastatic Tumor Tissue: Lung, 2x10 ug/PK
SDP
Supplier:  Biochain Institute Inc R8235152PM10
Encompass_Preferred
BioChain's human total RNA matched pair products include: Primary Pair (PP), Primary and Metastatic Pair (PM). PP consists of total RNA isolated from primary tumor and its adjacent normal tissue; PM consists of total RNA from primary tumor and and corresponding metastatic tumor. Total RNA in each pair is prepared from the same donor. This product line is designed for identifying tumor-specific genes and tumor metastatic genes.
Catalog No. 50-181-721