Learn More
Biochain Institute Inc Matched Pair - cDNA - Human Primary and Matched Metastatic Tumor Tissue: Liver, 10 x2 rxn /PK
SDP
Catalog No. 50181649
Encompass_Preferred
BioChain's human cDNA matched pair products include: Primary Pair (PP), Primary and Metastasis Pair (PM). PP consists of cDNA prepared from primary tumor and its adjacent normal tissue; PM consists of cDNA prepared from primary tumor and corresponding metastatic tumor. cDNAs in each pair are prepared from the same donor. This product line is designed for identifying tumor-specific (or disease specific) genes and tumor metastatic genes.
Catalog No. 50-181-649 Supplier Biochain Institute Inc Supplier No. C8235149PM
BioChain's human cDNA matched pair products include: Primary Pair (PP), Primary and Metastasis Pair (PM). PP consists of cDNA prepared from primary tumor and its adjacent normal tissue; PM consists of cDNA prepared from primary tumor and corresponding metastatic tumor. cDNAs in each pair are prepared from the same donor. This product line is designed for identifying tumor-specific (or disease specific) genes and tumor metastatic genes.