Learn More
Biochain Institute Inc Matched Pair - Paraffin Tissue Section - Human Primary Tumor and Normal (PP): Breast, 5 x2 slides /PK
SDP
Catalog No. 50181686
Encompass_Preferred
BioChain's human paraffin tissue section matched pair products include: Primary Pair (PP), Primary and Metastatic Pair (PM). PP consists of paraffin tissue sections of primary tumor and its adjacent normal tissue; PM consists of paraffin tissue sections of primary tumor and corresponding metastatic tumor. Paraffin tissue sections in each pair are prepared from the same donor. This product line is ideal for localization of tumor- specific genes and tumor metastatic genes.
Catalog No. 50-181-686 Supplier Biochain Institute Inc Supplier No. T8235086PP
BioChain's human paraffin tissue section matched pair products include: Primary Pair (PP), Primary and Metastatic Pair (PM). PP consists of paraffin tissue sections of primary tumor and its adjacent normal tissue; PM consists of paraffin tissue sections of primary tumor and corresponding metastatic tumor. Paraffin tissue sections in each pair are prepared from the same donor. This product line is ideal for localization of tumor- specific genes and tumor metastatic genes.