Learn More
Biochain Institute Inc Matched Pair - DNA - Human Primary and Metastatic Tumor Tissue: Rectum, 2x10 ug/PK
SDP
Supplier:  Biochain Institute Inc D8235206PM10
Encompass_Preferred
BioChain's human DNA matched pair products include: Primary Pair (PP), Primary and Metastasis Pair (PM). PP consists of DNA prepared from primary tumor and its adjacent normal tissue; PM consists of DNA prepared from primary tumor and corresponding ponding metastatic tumor. Genomic DNA in each pair is prepared from the same donor. This product line is designed foridentifying tumor-specific genes and tumor metastatic genes.
Catalog No. 50-181-667