Learn More
Biochain Institute Inc Matched Pair - Paraffin Tissue Section - Human Primary and Matched Metastatic Tumor (PM): Rectum, 5 x2 slides /PK
SDP
Catalog No. 50181695
Encompass_Preferred
BioChain's human paraffin tissue (FFPE) section matched pair products include: Primary Pair (PP), Primary and Metastatic Pair (PM). PP consists of paraffin tissue sections of primary tumor and its adjacent normal tissue; PM consists of paraffin tissue sections of primary tumor and corresponding metastatic tumor. Paraffin tissue sections in each pair are prepared from the same donor. This product line is ideal for localization of tumor- specific genes and tumor metastatic genes.
Catalog No. 50-181-695 Supplier Biochain Institute Inc Supplier No. T8235206PM
Add to Cart
Add to Cart
BioChain's human paraffin tissue (FFPE) section matched pair products include: Primary Pair (PP), Primary and Metastatic Pair (PM). PP consists of paraffin tissue sections of primary tumor and its adjacent normal tissue; PM consists of paraffin tissue sections of primary tumor and corresponding metastatic tumor. Paraffin tissue sections in each pair are prepared from the same donor. This product line is ideal for localization of tumor- specific genes and tumor metastatic genes.