Learn More
Biochain Institute Inc Matched Pair - Paraffin Tissue Section - Human Primary Tumor and Normal (PP): Liver, 5 x2 slides /PK
SDP
Supplier:  Biochain Institute Inc T8235149PP
Encompass_Preferred
BioChain's human paraffin tissue (FFPE) section matched pair products include: Primary Pair (PP), Primary and Metastatic Pair (PM). PP consists of paraffin tissue sections of primary tumor and its adjacent normal tissue; PM consists of paraffin tissue sections of primary tumor and corresponding metastatic tumor. Paraffin tissue sections in each pair are prepared from the same donor. This product line is ideal for localization of tumor- specific genes and tumor metastatic genes.
Catalog No. 50-181-692