Learn More
MSC Suburban Tool V-Block, Steel, 90°, 4-3/8 in. Capacity, 1/Pr.
Catalog No. 5000516916
Encompass
These matched 90° V-blocks provide a quick, accurate, and positive means of holding workpieces for layout, machining, grinding, and inspection tasks. Constructed from solid, one-piece steel and hardened to Rc 58-60, these V-blocks ensure durability and precision in demanding industrial environments.
  • Hardened to Rc 58-60 for extended service life
  • Matched pair precision within 0.0003 in.
  • Square, parallel, and central accuracy of 0.0005 in.
  • Maximum work holding capacity of 4-3/8 in. diameter
  • Includes one reversible, swing-away clamp for each block
  • Integrated side slots for easy mounting to work surfaces
  • Compatible with specific sine plate systems
Catalog No. 50-005-16916 Supplier MSC Supplier No. 05740048
These matched 90° V-blocks provide a quick, accurate, and positive means of holding workpieces for layout, machining, grinding, and inspection tasks. Constructed from solid, one-piece steel and hardened to Rc 58-60, these V-blocks ensure durability and precision in demanding industrial environments.
  • Hardened to Rc 58-60 for extended service life
  • Matched pair precision within 0.0003 in.
  • Square, parallel, and central accuracy of 0.0005 in.
  • Maximum work holding capacity of 4-3/8 in. diameter
  • Includes one reversible, swing-away clamp for each block
  • Integrated side slots for easy mounting to work surfaces
  • Compatible with specific sine plate systems