Learn More
Description
- Students will analyze simulated blood samples using real world techniques utilized in biomedical labs.
- Learn to count white blood cells using a hemocytometer.
- Perform blood compatibility tests to reveal information about donor blood samples.
- Determine who is the best candidate to donate a kidney to a patient in need.
- For 10 Lab Groups.
- Includes : Instructions, Control Simulated Blood Samples, Anti-A Serum, Anti-B Serum, Anti-Rh Serum, AHG Serum, Red Dye Concentrate, Simulated White Blood Cell Counting Sample, Transfer Pipets, Microtiter Plates, Microcentrifuge Tubes, Counting Chambers.
Specifications
Specifications
| Grade Level | High School, Higher Ed |
| Grade | 9 to 12 |
| Time Range | 60 Min. |
| Class Size | 10 Lab Groups |
| Product Type | ABO-Rh Blood Typing Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

