Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Material Dispenser with Liner Remover

Catalog No. 19129291 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-129-291 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-291 Supplier Brady™ Supplier No. DISPENSER
  • $666.00
    $448.65 / Each of 1

    Save $217.35

    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Don't rock the boat – weighted base and rubber feet keep the dispenser in place during on-the-go labeling projects

  • Save space with the narrow footprint
  • Innovative protective cover protects your material adhesive from exposure to the elements so your labels last
  • Your one-off identification tasks are quick and uncomplicated – keep it that way with the Brady Material Dispenser with Liner Remover
  • Specifications

    Product Type Label Dispenser
    Color Blue
    Height (Metric) 169 mm
    Material Steel
    For Use With (Application) Brady’s complete 3 in. core continuous material
    Quantity 1 Ea.
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.