Learn More
MSC SnowEx Optimum Flow Kit, Improves Material Flow, Vibrator, Dual Pitch Baffle
Catalog No. 502839402
Encompass
The SnowEx Optimum Flow Kit is designed to enhance the performance of V-Pro Poly Hopper Spreaders by significantly improving material flow. This kit incorporates a vibrator and a dual pitch baffle, working in conjunction to ensure more efficient and consistent spreading.
  • Improves material flow
  • Ensures improved spreading
  • Includes a vibrator
  • Includes a dual pitch baffle
Catalog No. 50-283-9402 Supplier MSC Supplier No. 57467607
The SnowEx Optimum Flow Kit is designed to enhance the performance of V-Pro Poly Hopper Spreaders by significantly improving material flow. This kit incorporates a vibrator and a dual pitch baffle, working in conjunction to ensure more efficient and consistent spreading.
  • Improves material flow
  • Ensures improved spreading
  • Includes a vibrator
  • Includes a dual pitch baffle