Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wanzl Material Handling Bulk Transport Cart

Catalog No. 11222182
Change view
Click to view available options
Color:
Chrome
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11-222-182 Chrome
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-222-182 Supplier Wanzl Supplier No. T.FC18376GY

missing translation for 'validMainframeMsg'

Bulk Transport Cart is a versatile method for transporting bulk materials. Center casters create cantilever action, promoting easier transport of heavy loads.

  • Ideal for transporting bulk materials
  • Center casters create a cantilever action, promoting easier handling of heavy loads
  • Made of Tubular Steel
  • (4) 5 in. Swivel Casters
  • (2) 5 in. Rigid/Fixed Center Casters

Specifications

Product Type Cart
Dimensions (D x W x H) 18 x 44 x 37 in.
Color Chrome
For Use With (Application) Transporting Bulk Materials
Material Steel (18 ga.)
No. of Casters 6
No. of Shelves 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.