Learn More
Motic™ Material Science Package: PX43MET Trino Inverted Microscope w/ Moticam S6 and Image-Pro Materials (2D) Software
Catalog No. 01673116 Shop All Motic Group Products
Change view
Click to view available options
Head Style:
Trinocular
Catalog No. Head Style
01-673-116 Trinocular
1 options
Catalog No. 01-673-116 Supplier Motic™ Supplier No. MBPX43MS6MAT1
Add to Cart
Add to Cart
Request a Quote
This Material Science Package includes: PX43MET Trinocular Inverted Microscope, Moticam S6 USB Camera, Image-Pro - Materials Package for Motic (2D), 0.5X C-Mount camera adapter, Polarizer, Analyzer
  • Microscope: PX43MET Trinocular Inverted Microscope The PX43MET is a professional Inverted Metallurgical microscope designed to inspect polished cross-sections as well as larger, bulkier materials samples.
  • The microscope is a best fit for automotive, machinery, and metal fabrication industries and similar fields. Digital Camera: Moticam S6 USB Camera The Hi-Res Partner - Moticam S6 is designed for those application fields which need higher resolution.
  • Presentations to a greater audience through video projector and reports in hardcopy are performed with ease.
  • With 6 million pixels and a fast data readout, the performance is remarkable! Software: Image-Pro - Materials Package for Motic (2D) Enhance your materials research with the Image-Pro - Materials Package for Motic, developed in partnership with Media Cybernetics.
  • This comprehensive solution integrates advanced Machine Learning technology with a detailed set of protocols to provide precise segmentation, classification, and automated measurements from 2D images. Designed for diverse materials applications, this package boosts your analytical capabilities with efficiency and accuracy.
  • Key Features:
  • Machine Learning-Powered Analysis Leverage sophisticated Machine Learning algorithms for accurate segmentation and classification, ensuring automated and reliable measurements from 2D images.
  • Materials Protocol Collection Streamline your research with an extensive suite of protocols ensuring efficient and repeatable analysis processes.
  • Designed for various materials applications, this collection includes protocols for Composite Materials, Fiber Separation, Fiber Thickness, Grain, Layers, Particles Phase, Particle Size, Phases, and Pores.
  • Efficiency and Accuracy Achieve high precision in your materials research with automated processes and detailed measurement capabilities.
  • Versatility Suitable for a wide range of materials science applications, making it an essential tool for researchers in diverse fields.
  • This package also includes a 12-month subscription to a Standard Success Plan from Media Cybernetics, providing ongoing technical assistance and software onboarding so you can focus on the work that truly matters.
Specifications
Voltage 115/230 VAC
Magnification Power 5X, 10X, 20X, 50X
Objective Types Plan S-APO BD
Software Type Image-Pro Materials (2D) Software
Eyepiece UV-WF 10X/FN22MM
Lighting Type LED, Transmitted Illumination
Head Style Trinocular
Camera Type Moticam S6
Display Type Digital
Model PX43MET
Product Type Inverted Trinocular Microscope