Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC MATHESON 3450 SERIES LINE REG
Catalog No. NC1759153
Encompass_Preferred
Matheson Model 3450 Series High-Purity/High Flow Stainless Steel Line Regulator, 1/4in FNPT connections, 4-100 psig delivery pressure range, 316 SS body, trim, diaphram, teflon seat, .6 Cv, outlet gauge 30in vac-0-200 psig, cleaned for O2 service
Catalog No. NC1759153 Supplier ATLANTIC BIOPHARM SOLUTIONS LLC Supplier No. SEQ3454S
Matheson Model 3450 Series High-Purity/High Flow Stainless Steel Line Regulator, 1/4in FNPT connections, 4-100 psig delivery pressure range, 316 SS body, trim, diaphram, teflon seat, .6 Cv, outlet gauge 30in vac-0-200 psig, cleaned for O2 service