Learn More
MSC Miller Full-Body Harness, Stretch Elastomer Webbing, Small, Mating Buckles, 1 ea.
Catalog No. 5000507730
Encompass
This full-body harness is constructed with green stretch elastomer webbing designed to adapt to the worker's shape, providing flexibility and movement while reducing fatigue. It is engineered for general purpose applications and incorporates features to enhance comfort, support, and productivity, with a configuration specifically designed to improve fit for female workers.
  • Features stretchable webbing to prevent bunching, binding, or kinking
  • Equipped with mating buckle chest and leg straps and friction shoulder buckles
  • Includes a sliding back D-ring, side D-rings, and a front positioning loop
  • Integrated back and leg pads for improved hip support and comfort
  • Hardware meets ASTM 50-hour salt spray test requirements
  • Minimum tensile strength of 6,000 lbs for webbing, 5,000 lbs for D-rings, and 4,000 lbs for buckles
  • Certified to applicable OSHA, ANSI, and CSA standards
Catalog No. 50-005-07730 Supplier MSC Supplier No. 91515361
This full-body harness is constructed with green stretch elastomer webbing designed to adapt to the worker's shape, providing flexibility and movement while reducing fatigue. It is engineered for general purpose applications and incorporates features to enhance comfort, support, and productivity, with a configuration specifically designed to improve fit for female workers.
  • Features stretchable webbing to prevent bunching, binding, or kinking
  • Equipped with mating buckle chest and leg straps and friction shoulder buckles
  • Includes a sliding back D-ring, side D-rings, and a front positioning loop
  • Integrated back and leg pads for improved hip support and comfort
  • Hardware meets ASTM 50-hour salt spray test requirements
  • Minimum tensile strength of 6,000 lbs for webbing, 5,000 lbs for D-rings, and 4,000 lbs for buckles
  • Certified to applicable OSHA, ANSI, and CSA standards